Skip to Content
Merck
All Photos(1)

Documents

BR701450

BRAND® Reagent Reservoirs for multichannel pipettes and automatic systems

1 column, standard profile, column capacity 220 mL, pyramid bottom (96-well)

Synonym(s):

ldpe bottle, polyethylene bottles, polyethylene containers

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121522
NACRES:
NB.01

material

colorless
polypropylene
pyramid bottom (96-well)

sterility

non-sterile

feature

RNase and DNase free
1 column
standard profile

packaging

pack of 10

manufacturer/tradename

BRAND 701450

capacity

220 mL

column capacity

220 mL

Looking for similar products? Visit Product Comparison Guide

General description

These reagent reservoirs were designed especially for use with automated liquid handling platforms. They can also be used with manual systems. Different partitions, bottom styles and profiles provide great application flexibility.

Features and Benefits

  • ANSI/SLAS standard format Compatible with nearly all liquid handling robots and other automated dispensing systems
  • 5 different types for use with multichannel pipettes and automatic systems
  • Excellent stability and chemical resistance made from high-quality polypropylene
  • Very low dead volume due to pyramid bottoms
  • Free from RNases and DNases
  • Autoclavable (121 °C, 20 min)

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service