Skip to Content
Merck
All Photos(5)

Documents

AV34094

Sigma-Aldrich

Anti-NPM1 (AB1) antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Nucleophosmin (nucleolar phosphoprotein B23, numatrin)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

33 kDa

species reactivity

mouse, dog, pig, human, horse, rabbit, rat

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NPM1(4869)

Immunogen

Synthetic peptide directed towards the N terminal region of human NPM1

Application

Mouse retinas were subjected to western blot analysis using rabbit anti-NPM1 with incubation overnight at 4 degrees. Antibody was diluted 1:1000.

Biochem/physiol Actions

NPM1 is a member of Nucleophosmin (NPM) family. NPM is a ubiquitously expressed nucleolar phosphoprotein that continuously shuttles between the nucleus and cytoplasm

Sequence

Synthetic peptide located within the following region: DMDMSPLRPQNYLFGCELKADKDYHFKVDNDENEHQLSLRTVSLGAGAKD

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrey Turchinovich et al.
Nucleic acids research, 39(16), 7223-7233 (2011-05-26)
MicroRNAs (miRNAs), a class of post-transcriptional gene expression regulators, have recently been detected in human body fluids, including peripheral blood plasma as extracellular nuclease resistant entities. However, the origin and function of extracellular circulating miRNA remain essentially unknown. Here, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service