Skip to Content
Merck
All Photos(1)

Key Documents

A6854

Sigma-Aldrich

AF12198

≥98% (HPLC), solid

Synonym(s):

Ac-Phe-Glu-Trp-Thr-Pro-Gly-Trp-Tyr-Gln-L-azetidine-2-carbonyl-Tyr-Ala-Leu-Pro-Leu-NH2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C96H123N19O22
CAS Number:
Molecular Weight:
1895.12
MDL number:
UNSPSC Code:
51111800

Assay

≥98% (HPLC)

form

solid

mol wt

1895.12

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... PNOC(5368)

Amino Acid Sequence

Ac-Phe-Glu-Trp-Thr-Pro-Gly-Trp-Tyr-Gln-Aze-Tyr-Ala-Leu-Pro-Leu-NH2

Biochem/physiol Actions

Human IL-1 receptor antagonist peptide, in vivo and in vitro.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service