A1354
APRIL human
recombinant, expressed in NSO cells, >95% (SDS-PAGE), lyophilized powder
Synonym(s):
A Proliferation-inducing Ligand, TALL-2, TNFSF13, TRDL-1
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
MDL number:
UNSPSC Code:
12352200
recombinant
expressed in NSO cells
Assay
>95% (SDS-PAGE)
form
lyophilized powder
mol wt
23-25 kDa by SDS-PAGE (reducing)
calculated mol wt 18.6 kDa
impurities
endotoxin, tested
UniProt accession no.
storage temp.
−20°C
Gene Information
human ... TNFSF13(8741)
Physical form
Lyophilized from a 0.2 μm filtered solution in PBS and 0.5 M NaCl, containing 50 μg BSA per 1 μg cytokine
Analysis Note
The biological activity is measured by its ability to induce proliferation of anti-IgM-stimulated human B cells. Alternatively, the activity can be measured by its ability to bind immobilized rhBCMA/Fc.
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
K Kelly et al.
Cancer research, 60(4), 1021-1027 (2000-03-08)
We have examined the activity of a new member of the tumor necrosis factor (TNF) family identified through Expressed Sequence Tag database searching using TNFalpha protein as the search query. We have termed this protein TNF-related death ligand-la (TRDL-1alpha). Traditional
P A Moore et al.
Science (New York, N.Y.), 285(5425), 260-263 (1999-07-10)
The tumor necrosis factor (TNF) superfamily of cytokines includes both soluble and membrane-bound proteins that regulate immune responses. A member of the human TNF family, BLyS (B lymphocyte stimulator), was identified that induced B cell proliferation and immunoglobulin secretion. BLyS
M Hahne et al.
The Journal of experimental medicine, 188(6), 1185-1190 (1998-09-22)
Members of the tumor necrosis factor (TNF) family induce pleiotropic biological responses, including cell growth, differentiation, and even death. Here we describe a novel member of the TNF family designated APRIL (for a proliferation-inducing ligand). Although transcripts of APRIL are
J A Gross et al.
Nature, 404(6781), 995-999 (2000-05-09)
B cells are important in the development of autoimmune disorders by mechanisms involving dysregulated polyclonal B-cell activation, production of pathogenic antibodies, and co-stimulation of autoreactive T cells. zTNF4 (BLyS, BAFF, TALL-1, THANK) is a member of the tumour necrosis factor
P Schneider et al.
The Journal of experimental medicine, 189(11), 1747-1756 (1999-06-08)
Members of the tumor necrosis factor (TNF) family induce pleiotropic biological responses, including cell growth, differentiation, and even death. Here we describe a novel member of the TNF family, designated BAFF (for B cell activating factor belonging to the TNF
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service