Skip to Content
Merck
All Photos(1)

Key Documents

49511

Sigma-Aldrich

Chromeon 642-Streptavidin

BioReagent, suitable for fluorescence

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352108
NACRES:
NA.32

product line

BioReagent

form

solid

fluorescence

λex 642 nm; λem 665 nm in PBS

suitability

suitable for fluorescence

storage temp.

2-8°C

Application

Streptavidin-Chromeon 642 is used to label biotin-conjugated (biotinylated) molecules and biotin derivatized surfaces in a wide variety of immunofluorescence applications such as immunochemistry and histochemistry. Streptavidin- Chromeon 642 may be used in the development of bioassay devices such as Lab-on-a-chip (LoC) systems and microarrays.

Components

1 vial contains 1 mg pigment in 1 mL phosphate buffered saline pH 7.2, sodium azide 15 mmol/L

Legal Information

Licensed from Chromeon GmbH, Regensburg, Germany. Only for R&D use, reselling or repackaging not allowed. Use in any fluorescent lifetime assays requires a separate license.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Bruck et al.
Biosensors & bioelectronics, 26(9), 3832-3837 (2011-03-23)
We report the development of a Mach-Zehnder interferometer biosensor based on a high index contrast polymer material system and the demonstration of label-free online measurement of biotin-streptavidin binding on the sensor surface. The surface of the polyimide waveguide core layer

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service