114PR-500
Endothelial Cell Serum-Free, Growth Factor-Free Medium without Phenol Red (500 ml)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
form
liquid
Quality Level
shelf life
6 mo.
manufacturer/tradename
Cell Applications, Inc
shipped in
wet ice
storage temp.
2-8°C
General description
A proprietary medium developed to induce morphological differentiation of CADMEC™ in Attachment Factor Solution coated tissue culture ware in 50% confluent culture. This medium induces CADMEC™ to form cord-like structures in 48 hours. These differentiated cells will resume their cobblestone morphology when switched back to CADMEC™ Growth Medium. It is supplemented with defined components to allow angiogenesis studies on the Attachment Factor-coated tissue culture ware without the interference of undefined components in Matrigel.
Quality
Each lot was tested for the ability to support plating, spreading and proliferation
Preparation Note
contained in protocol
Legal Information
CADMEC is a trademark of Cell Applications
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service