Skip to Content
Merck
All Photos(1)

Key Documents

15327

Sigma-Aldrich

Lead(II) chromate

purum p.a., ACS reagent, ≥98.0% (RT)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
PbCrO4
CAS Number:
Molecular Weight:
323.19
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NB.24

grade

ACS reagent
purum p.a.

Assay

≥98.0% (RT)

form

powder

SMILES string

[PbH2++].[O-][Cr]([O-])(=O)=O

InChI

1S/Cr.4O.Pb/q;;;2*-1;+2

InChI key

MOUPNEIJQCETIW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Carc. 1A - Repr. 1A - STOT RE 2

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of sulphide in effluent streams using a solid-phase lead (II) chromate reactor incorporated into a flow-injection system
Van Staden JF and Kluever LG.
Analytica Chimica Acta, 369(1), 157-161 (1998)
Solid phase extraction of lead (II) by sorption on grinded eucalyptus stem and determination with flame atomic absorption spectrometry.
Mousavi HZ, et al.
J. Chin. Chem. Soc., 56(5), 974-974 (2009)
The electrokinetic potential of sparingly soluble salts.
Morimoto T.
Bulletin of the Chemical Society of Japan, 37(3), 386-392 (1964)
X I Gu et al.
Osteoarthritis and cartilage, 20(9), 1011-1019 (2012-05-23)
Recent developments on high resolution micro computed tomography (μCT) allow imaging of soft tissues in small animal joints. Nevertheless, μCT images cannot distinguish soft tissues from synovial fluid due to their similar mass density, limiting the 3D assessment of soft
Eric Camyre et al.
Toxicological sciences : an official journal of the Society of Toxicology, 97(2), 348-354 (2007-03-16)
Particulate hexavalent chromium ((Cr(VI)) compounds are human lung carcinogens. These compounds induce DNA damage, chromosome aberrations, and concentration-dependent cell death in human and Chinese hamster ovary (CHO) cells. The relationship between Cr(VI)-induced DNA damage and chromosome aberrations is poorly understood.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service