Skip to Content
Merck
All Photos(1)

Documents

STS0012

Sigma-Aldrich

ECOSURF EH-9 solution

greener alternative

90% aqueous solution

Synonym(s):

2-Ethyl hexanol EoPo copolymer (9 EO), Alcohol (2EH) alkoxylate, Ethyl hexanol ethoxylated propoxylated

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12161902
NACRES:
NA.21

grade

laboratory grade

Quality Level

description

non-ionic

form

liquid

greener alternative product characteristics

Design for Degradation
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

CMC

1066 ppm (25°C)

viscosity

61.8 cSt(40 °C)

transition temp

pour point -5 °C
cloud point 64 °C (10 wt% actives aq solution)

solubility

water: soluble

density

1.0157 g/mL at 40 °C

HLB

12.5

greener alternative category

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product is a biodegradable surfactant and is aligned with the 10th principle of Green Chemistry "Design for Degradation".

This product is a 90 % aqueous solution of an alcohol ethoxylate

Application


  • Polyolefin-Supported Hydrogels for Selective Cleaning Treatments of Paintings.: This study explores the use of polyolefin-supported hydrogels incorporating ECOSURF; EH-9 solution for the selective cleaning of paintings. The hydrogels demonstrate effective cleaning capabilities while preserving the integrity of the artwork, highlighting their potential application in art conservation (Freese et al., 2019) (Freese et al., 2019).

Features and Benefits

  • Superior wetting
  • Excellent oily soil removal
  • Rapid dissolution & good rinseability
  • Low odor
  • No gel range
  • Outstanding formulation & handling properties
  • Readily biodegradable
  • Aquatic toxicity EC50 > 10 mg/L

Legal Information

ECOSURF is a trademark of The Dow Chemical Company ("Dow") or an affiliated company of Dow

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service