Skip to Content
Merck
All Photos(7)

Documents

P7632

Sigma-Aldrich

Pararosaniline Base

crystalline

Synonym(s):

Parafuchsin, Paramagenta

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H19N3O
CAS Number:
Molecular Weight:
305.37
Colour Index Number:
42500
Beilstein:
2148632
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

product name

Pararosaniline Base, crystalline

form

crystalline

Quality Level

color

purple

mp

205 °C (dec.) (lit.)

solubility

ethanol: 1 mg/mL, clear to hazy

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Nc1ccc(cc1)C(O)(c2ccc(N)cc2)c3ccc(N)cc3

InChI

1S/C19H19N3O/c20-16-7-1-13(2-8-16)19(23,14-3-9-17(21)10-4-14)15-5-11-18(22)12-6-15/h1-12,23H,20-22H2

InChI key

KRVRUAYUNOQMOV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Davide Tamburro et al.
Journal of the American Chemical Society, 133(47), 19178-19188 (2011-10-18)
Many low-abundance biomarkers for early detection of cancer and other diseases are invisible to mass spectrometry because they exist in body fluids in very low concentrations, are masked by high-abundance proteins such as albumin and immunoglobulins, and are very labile.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service