Skip to Content
Merck
All Photos(1)

Documents

NIST1107

Naval brass UNS 46400

NIST® SRM® 1107

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of disk

manufacturer/tradename

NIST®

format

matrix material

General description

Each unit of SRM 1107 consists of a solitary disk measuring around 3.2 cm in diameter and 1.9 cm in thickness. For more information, please refer to the COA and SDS.
SRM 1107_cert
SRM 1107_SDS

Application

Standard reference material 1107 is intended for validating chemical and instrumental methods for elemental analysis of brass alloys.

Features and Benefits

  • Available with NIST-certified values as well as reference mass fraction and Coverage Factor (k) values of elements in Naval brass estimated using a Gaussian random effects model [3–5] and the DerSimonian-Laird procedure.
  • The certified values are metrologically traceable to the SI unit of mass, expressed as a percent.
  • Information values for elements are provided and expressed as mass fractions.
  • Expiration details and instructions for use, storage, and handling are provided on the NIST certificate.

Other Notes

Certified for the analytes listed below.
Copper (Cu), Iron (Fe), Lead (Pb), Nickel (Ni), Tin (Sn), Zinc (Zn)

Reference/Informational value is provided for the following analytes. Aluminium (Al), Manganese (Mn), Phosphorus (P)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service