Skip to Content
Merck
All Photos(1)

Documents

BCR341

Benzo[b]naphtho[2,1-d]furan

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H10O
CAS Number:
Molecular Weight:
218.25
Beilstein:
156008
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

storage temp.

2-8°C

SMILES string

c1ccc2c(c1)ccc3c4ccccc4oc23

InChI

1S/C16H10O/c1-2-6-12-11(5-1)9-10-14-13-7-3-4-8-15(13)17-16(12)14/h1-10H

InChI key

BCBSVZISIWCHFM-UHFFFAOYSA-N

Analysis Note

For more information please see:
BCR341

Legal Information

BCR is a registered trademark of European Commission

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service