Skip to Content
Merck
All Photos(2)

Documents

908800

Sigma-Aldrich

Chiralyst Rh660

Umicore

Synonym(s):

1,2-Bis[(2R,5R)-2,5-diethylphospholano]benzene(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate, (R,R)-Et-DUPHOS-Rh, [(R,R)Ethyl-DuPhosRh(COD)]BF4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H48BF4P2Rh
CAS Number:
Molecular Weight:
660.36
UNSPSC Code:
12352005
NACRES:
NA.22

form

powder

SMILES string

C[CH+]C.CC[C@@H]1CC[C@@H](CC)P12C3=C(P4([Rh](C)2C)[C@H](CC)CC[C@H]4CC)C=CC=C3.FB(F)F.C5C/C=C\CC/C=C\5.[F-]

InChI

1S/C22H36P2.C8H12.BF4.Rh/c1-5-17-13-14-18(6-2)23(17)21-11-9-10-12-22(21)24-19(7-3)15-16-20(24)8-4;1-2-4-6-8-7-5-3-1;2-1(3,4)5;/h9-12,17-20H,5-8,13-16H2,1-4H3;1-2,7-8H,3-6H2;;/q;;-1;+1/b;2-1-,8-7-;;/t17-,18-,19-,20-;;;/m1.../s1

InChI key

DSYBEQKPSKLNMC-KYOOHHHUSA-N

Looking for similar products? Visit Product Comparison Guide

Application

DuPhos and BPE Ligands: Highly Efficient Privileged Ligands

Catalyst for:
  • Asymmetric hydrogenation reactions
Chiralyst Rh660 is a homogeneous catalyst useful for asymmetric reactions and asymmetric hydrogenation.
The catalyst works well with privileged ligands.

Product can be used with our benchtop hydrogen generator, H-Genie Lite (Z744083)

Storage and Stability

Do not store above 25°C (77°F). Store in a tightly closed original container under dry inert gas, protected from direct sunlight in a dry, cool and well-ventilated area.

Legal Information

Product of Umicore

Additional information available at www.pmc.umicore.com

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service