Skip to Content
Merck
All Photos(1)

Documents

68985

Supelco

4-tert-Amylphenol

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5C(CH3)2C6H4OH
CAS Number:
Molecular Weight:
164.24
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

255 °C (lit.)

mp

88-89 °C (lit.)

application(s)

environmental

format

neat

SMILES string

CCC(C)(C)c1ccc(O)cc1

InChI

1S/C11H16O/c1-4-11(2,3)9-5-7-10(12)8-6-9/h5-8,12H,4H2,1-3H3

InChI key

NRZWYNLTFLDQQX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-tert-Amylphenol belongs to the class of alkylphenols (APs). APs are predominantly utilized in various agricultural and personal care products. 4-tert-amylphenol (4-TAP) is used in the manufacture of fragrances, de-emulsifying agents, and biocides. It is an endocrine-disrupting chemical and is regarded as a persistent toxic environmental contaminant.

Application

4-tert-Amylphenol may be used as an analytical reference standard for the determination of the analyte in environmental samples by chromatography-based techniques.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of a fabric phase sorptive extraction with high-performance liquid chromatography and ultraviolet detection method for the analysis of alkyl phenols in environmental samples
Kumar R, et al.
Journal of Separation Science, 38(18), 3228-3238 (2015)
Analysis of multiple endocrine disruptors in environmental waters via wide-spectrum solid-phase extraction and dual-polarity ionization LC-ion trap-MS/MS
Benijts T, et al.
Analytical Chemistry, 76(3), 704-711 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service