Skip to Content
Merck
All Photos(1)

Key Documents

59428

Sigma-Aldrich

N-Isopropylhydroxylamine hydrochloride

≥97.0% (AT)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHNHOH · HCl
CAS Number:
Molecular Weight:
111.57
Beilstein:
3666413
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥97.0% (AT)

form

solid

storage temp.

−20°C

SMILES string

Cl.CC(C)NO

InChI

1S/C3H9NO.ClH/c1-3(2)4-5;/h3-5H,1-2H3;1H

InChI key

BYXUIKZQGOPKFR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chen Liu et al.
Cell research, 31(10), 1106-1122 (2021-07-10)
Whereas the critical roles of innate lymphoid cells (ILCs) in adult are increasingly appreciated, their developmental hierarchy in early human fetus remains largely elusive. In this study, we sorted human hematopoietic stem/progenitor cells, lymphoid progenitors, putative ILC progenitor/precursors and mature
Lin Yi et al.
Analytical and bioanalytical chemistry, 386(3), 666-674 (2006-05-26)
Characterizing the biological effects of metabolic transformations (or biotransformation) is one of the key steps in developing safe and effective pharmaceuticals. Sulfate conjugation, one of the major phase II biotransformations, is the focus of this study. While this biotransformation typically

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service