Skip to Content
Merck
All Photos(1)

Key Documents

45577

Supelco

Metiram

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116107

grade

analytical standard

description

technical mixture

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

−20°C

InChI

1S/C4H8N2S4.Zn/c7-3(8)5-1-2-6-4(9)10;/h1-2H2,(H2,5,7,8)(H2,6,9,10);/q;+2/p-2

InChI key

AMHNZOICSMBGDH-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Kornuta et al.
Journal of environmental pathology, toxicology and oncology : official organ of the International Society for Environmental Toxicology and Cancer, 15(2-4), 75-78 (1996-01-01)
Epidemiologic data showed an increase in the number of cancer cases in persons involved in agricultural production using pesticides. According to IARC, more than 25% of pesticides are classified as oncogens. In recent years, the concept of malignant tumors developing
[Hygienic characteristics of the working conditions in manufacturing polycarbacin (industrial and experimental research)].
V R Kokot
Gigiena truda i professional'nye zabolevaniia, (2)(2), 33-36 (1988-02-01)
L V Grimaĭlo et al.
Meditsinskaia parazitologiia i parazitarnye bolezni, (3)(3), 23-25 (1993-05-01)
Polycarbocin-lupine and polycarbocin-pea systems were found to exert a marked ovicidal effect providing decontamination of the top soil layer from ascarid eggs for 3 months. A combination of 4-5 mg of polycarbacine and 25-30 lupine or 20-30 pea plants per
[Toxicologic characteristics of combined pesticides on the base polycarbacin].
M I Gzhegotskiĭ et al.
Gigiena truda i professional'nye zabolevaniia, (8)(8), 51-52 (1987-08-01)
[Determination of maximum permissible concentration of polycarbacide in the water reservoirs].
K K Brochinskiĭ et al.
Gigiena i sanitariia, (11)(11), 76-77 (1980-11-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service