Skip to Content
Merck
All Photos(1)

Key Documents

37349

Sigma-Aldrich

9,10-Dihydrophenanthrene

technical, ≥90% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H12
CAS Number:
Molecular Weight:
180.25
Beilstein:
1907655
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

technical

Assay

≥90% (GC)

bp

168-169 °C/15 mmHg (lit.)

mp

30-35 °C (lit.)
30-35 °C

SMILES string

C1Cc2ccccc2-c3ccccc13

InChI

1S/C14H12/c1-3-7-13-11(5-1)9-10-12-6-2-4-8-14(12)13/h1-8H,9-10H2

InChI key

XXPBFNVKTVJZKF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service