33945
Malachite Green-d5 picrate
VETRANAL®, analytical standard
Synonym(s):
Bis-(4-dimethylaminophenyl)-phenyl-d5-carbenium picrate
About This Item
Recommended Products
grade
analytical standard
Quality Level
product line
VETRANAL®
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
cleaning products
cosmetics
environmental
food and beverages
personal care
format
neat
mass shift
M+5
SMILES string
[O-]c1c(cc(cc1[N+]([O-])=O)[N+]([O-])=O)[N+]([O-])=O.[2H]c2c([2H])c([2H])c(c([2H])c2[2H])\C(c3ccc(cc3)N(C)C)=C4\C=CC(\C=C4)=[N+](/C)C
InChI
1S/C23H25N2.C6H3N3O7/c1-24(2)21-14-10-19(11-15-21)23(18-8-6-5-7-9-18)20-12-16-22(17-13-20)25(3)4;10-6-4(8(13)14)1-3(7(11)12)2-5(6)9(15)16/h5-17H,1-4H3;1-2,10H/q+1;/p-1/i5D,6D,7D,8D,9D;
InChI key
HKJSJDRORUIJJI-AYVDECCJSA-M
General description
Application
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Expl. 1.1
Storage Class Code
1 - Explosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service