ZFCBIXXIT
Country Box Milli-Q® IX Systems
Availabe only in Italy
Synonym(s):
Country Box Milli-Q® IX IT
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
packaging
pkg of 1 kit
availability
available only in Italy
compatibility
for use with Milli-Q® IX 7003
for use with Milli-Q® IX 7005
for use with Milli-Q® IX 7010
for use with Milli-Q® IX 7015
Looking for similar products? Visit Product Comparison Guide
General description
The Country Box for Milli-Q® IX 7003/05/10/15 pure water systems is an installation accessory that contains a printed Quick Reference Guide, a user manual in local language and in English on a USB key, and a power cord adapted to the local electrical network.
Application
The components of the country box assist users to quickly access the information they need to correctly use their system and connect to the local power network.
Legal Information
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service