Skip to Content
Merck
All Photos(1)

Key Documents

MMHEADMIC

Millipore

Milliflex Oasis® Filtration Head

for use with Microfil® filtration units

Synonym(s):

Filtration Head, Manifold Head, Milliflex Oasis® Pump Head, Milliflex Oasis® filtration pump head

Sign Into View Organizational & Contract Pricing


About This Item

NACRES:
SB.56
Pricing and availability is not currently available.

material

316L stainless steel (ASI)

Sterilization Compatibility

autoclavable compatible

packaging

pkg of 1 unit

manufacturer/tradename

Milliflex Oasis®

application(s)

bioburden testing
food and beverages
pharmaceutical
water monitoring

compatibility

for use with Microfil® filtration units

shipped in

ambient

General description

Milliflex Oasis® Pump head is designed to support during filtration process. The filtration head is compatible for Microfil® funnel units. Milliflex Oasis®Pump head facilitate high throughput filtration for bioburden and water quality testing.

Application

Milliflex Oasis® Pump head is used to fit Microfil® funnel units for filtration process carried out to determine bioburden load and water quality.

Legal Information

MICROFIL is a registered trademark of Merck KGaA, Darmstadt, Germany
MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
MILLIFLEX OASIS is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service