Skip to Content
Merck
All Photos(1)

Key Documents

HPI-15K

Sigma-Aldrich

Human Proinsulin RIA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.84
Pricing and Inventory is not available. Please check the Merck website for more information.

grade

for protein analysis

Quality Level

description

ED<sub>80</sub> = 10 ± 1 pM ED<sub>50</sub> = 23 ± 2 pM ED<sub>20</sub> = 52 ± 3 pM
analytes available: Proinsulin
2-100 pg/mL (standard curve range)

feature

accuracy 91–93%

species reactivity

human

technique(s)

radioimmunoassay: suitable

UniProt accession no.

detection method

radioactive

General description

FUNCTION: SwissProt: P01308 # Insulin decreases blood glucose concentration. It increases cell permeability to monosaccharides, amino acids and fatty acids. It accelerates glycolysis, the pentose phosphate cycle, and glycogen synthesis in liver.
SIZE: 110 amino acids; 11981 Da
SUBUNIT: Heterodimer of a B chain and an A chain linked by two disulfide bonds.
SUBCELLULAR LOCATION: Secreted.
DISEASE:SwissProt: P01308 # Defects in INS are the cause of familial hyperproinsulinemia [MIM:176730].
SIMILARITY: SwissProt: P01308 ## Belongs to the insulin family.

Application

Three-day, disequilibrium incubation at room temperature. Sample volume ≤200 µL serum, plasma, or tissue culture media.

Analysis Note

Linearity 100–120%

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - STOT RE 2

Target Organs

Respiratory Tract

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service