Skip to Content
Merck
All Photos(1)

Documents

GF39

Sigma-Aldrich

Anti-HGF (Scatter Factor) (Ab-1) Mouse mAb (4C12.1)

liquid, clone 4C12.1, Calbiochem®

Synonym(s):

Anti-Hepatocyte Growth Factor, Anti-SF, Anti-Hepatopoeitin A, Anti-Scatter Factor, Anti-Scatter Factor, Anti-Hepatocyte Growth Factor, Anti-SF, Anti-Hepatopoeitin A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

4C12.1, monoclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

isotype

IgG2b

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... HGF(3082)

General description

Anti-HGF (Scatter Factor) (Ab-1), mouse monoclonal, clone 4C12.1, recognizes the ~82 kDa hepatocyte growth factor in macrophages, endothelial & Kupffer cells. It is validated for use in WB & IP.
Purified mouse monoclonal antibody. Recognizes the ~82 kDa HGF protein.
Recognizes the ~82 kDa hepatocyte growth factor protein in macrophages, endothelial, and Kupffer cells.

Immunogen

Human
recombinant, human HGF protein

Application

Immunoblotting (1:200-1:400)

Immunoprecipitation (10 µl/mg protein lysate)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 10 mM PBS, 0.2% BSA, pH 7.4.

Reconstitution

Following initial use, aliquot and freeze (-20°C) for long-term storage if desired. Avoid freeze/thaw.

Analysis Note

Positive Control
Endothelial cells, Kupffer cells, or macrophages

Other Notes

Antibody should be titrated for optimal results in individual systems.
Toi, M., et al. 1998. Clin. Cancer Res.4, 659.
Bardelli, A. and Comoglio, P.M. 1997. Ciba Found. Symp.212, 133.
Birchmeier, C., et al. 1997. Ciba Found. Symp.212, 16.
Jiang, W.G. and Hiscox, S. 1997. Histol. Histopathol.12, 537.
Matsumoto, K. and Nakamura, T. 1997. Ciba Found. Symp.212, 198.
Rosen, E.M., et al. 1997. Ciba Found. Symp.212, 215.
Vande Woude, G.F., et al. 1997. Ciba Found Symp.212, 119.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service