Skip to Content
Merck
All Photos(3)

Documents

ABE411

Sigma-Aldrich

Anti-dimethyl Histone H3 (Arg26), Asymmetric Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Histone H3.1, Histone H3/a, Histone H3/b, Histone H3/c, Histone H3/d, Histone H3/f, Histone H3/h, Histone H3/i, Histone H3/j, Histone H3/k, Histone H3/l

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

dot blot: suitable
immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

dimethylation (Arg26)

Gene Information

human ... HIST1H3F(8968)

General description

Histone H3 is one of the five main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N-terminal tail, H3 is involved with the structure of the nucleosomes of the ′beads on a string′ structure. The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of epigenetic modifications that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine.

Specificity

Broad species reactivity is expected based on immunogen sequence similarities.

Application

Immunocytochemistry Analysis: A 1:500 dilution from a representative lot detected dimethyl Histone H3 (Arg26), Asymmetric in A431 and HepG2 cells.

Dot Blot (Specificity) Analysis: 0.5 µg/mL from a representative lot detected dimethyl Histone H3 (Arg26), Asymmetric in Absurance Histone H3 Antibody Specificity Array (Cat. No. 16-667) and Absurance Histone H2A, H2B, H4 Antibody Specificity Array (Cat. No. 16-665).

DyLight is a registered trademark of Thermo Fisher Scientific.
This Anti-dimethyl Histone H3 (Arg26) antibody, Asymmetric is validated for use in western blotting, ICC & dot blot for the detection of dimethyl Histone H3 (Arg26).

Quality

Evaluated by Western Blotting in untreated and CARM1 treated HeLa acid extracts.

Western Blotting Analysis: 1 µg/mL of this antibody detects dimethyl Histone H3 (Arg26), Asymmetric in 10 µg of CARM1 treated HeLa acid extract.

Target description

~17 kDa observed

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service