Skip to Content
Merck
All Photos(1)

Documents

ABE2915

Sigma-Aldrich

Anti-MyT1L

Synonym(s):

MyT1-L, Myelin transcription factor 1-like protein, NZF-1, Neural zinc finger factor 1, Postmitotic neural gene 1 protein, Zinc finger protein Png-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

mol wt

calculated mol wt 132.95 kDa
observed mol wt ~170 kDa (Uncharacterized bands may be observed in some lysate(s).)

species reactivity

mouse

isotype

IgG

shipped in

ambient

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Myt1L(17933)

Specificity

This rabbit polyclonal antibody specifically detects murine Myelin transcription factor 1-like protein. It targets an epitope within the N-terminal half.

Immunogen

His-tagged recombinant fragment corresponding to 250 amino acids from the N-terminal half of mouse Myelin transcription factor 1-like protein

Target description

Myelin transcription factor 1-like protein (UniProt: P97500; also known as MyT1-L, MyT1L, Neural zinc finger factor 1, NZF-1, Postmitotic neural gene 1 protein, Zinc finger protein Png-1) is encoded by the Myt1l (also known as Kiaa1106, Nzf1, Png1) gene (Gene ID: 17933) in murine species. MyT1l is a pan neuron-specific transcription factor that plays a key role in neuronal differentiation by specifically repressing expression of non-neuronal genes during neuron differentiation. It preferentially binds to DNA binding sites that are in an open chromatin configuration. It is reported to exert its pro-neuronal function by direct repression of many different somatic lineage programs. MyT1l inhibits specific lineages and mediates repression of multiple differentiation programs. Its repressive function is mediated via recruitment of a complex containing Sin3b by binding to N-terminal domain. It is also shown to repress the expression of negative regulators of neurogenesis, such as members of the Notch signaling pathways, including Hes1. The combination of ASCL1, POU3F2/BRN2, and MyT1L is shown to be sufficient to reprogram fibroblasts into induced neuronal (iN) cells and administration of exogenous Myt1l can greatly enhance the efficiency of reprogramming and the functional maturity of the resulting iN cells. (Ref.: Mall, M., et al. (2017). Nature. 544(7649); 245-249).

Physical form

Purified rabbit polyclonal antibody in PBS with 0.05% sodium azide with 50% glycerol.

Reconstitution

Please refer to lot specific datasheet.

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service