Skip to Content
Merck
All Photos(2)

Key Documents

ABE1956

Sigma-Aldrich

Anti-DNMT1 Antibody, Ac-4K

from rabbit

Synonym(s):

DNA (cytosine-5)-methyltransferase 1, EC: 2.1.1.37, CXXC-type zinc finger protein 9, DNA methyltransferase HsaI, DNA MTase HsaI, M.HsaI, MCMT

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

species reactivity (predicted by homology)

mouse (based on 100% sequence homology), rat (based on 100% sequence homology)

packaging

antibody small pack of 25 μg

technique(s)

dot blot: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... DNMT1(1786)

General description

DNA (cytosine-5)-methyltransferase 1 (UniProt: P26358; also known as EC: 2.1.1.37, Dnmt1, CXXC-type zinc finger protein 9, DNA methyltransferase HsaI, DNA MTase HsaI, M.HsaI, MCMT) is encoded by the DNMT1 (also known as AIM, CXXC9, DNMT) gene (Gene ID: 1786) in human. Dnmt1 is an important epigenetic regulator that plays a key role in the maintenance of DNA methylation. Dnmt1 is a homodimeric protein that is ubiquitous in its expression. It is highly expressed in fetal tissues and in heart, kidney, placenta and peripheral mononuclear cells. Its abundance is reduced to almost non-detectable levels at the G0 phase of the cell cycle and is dramatically induced upon entrance into the S-phase of the cell cycle. Its expression is regulated by many signaling pathways, including PI3/Akt, Rb/E2F and p53/SP1. Dnmt1 is shown to preferentially methylate hemimethylated DNA and it associates with DNA replication sites in the S phase to maintain the methylation pattern in newly synthesized strand, which is essential for epigenetic inheritance. Dnmt1 associates with chromatin during G2 and M phases to maintain DNA methylation independently of replication. It is also responsible for maintaining methylation patterns established in development. Dnmt1 can undergo acetylation on multiple lysines mainly by KAT2B/PCAF. It can be phosphorylated on Serine 154 by Cdks and this phosphorylation is considered to be essential for its enzymatic activity and protein stability. Its phosphorylation on Serine 143 by Akt1 is reported to prevent methylation by SETD7, which increases its stability. Mutations in DNMT1 gene are reported to cause hereditary sensory neuropathy that is characterized by adult onset of progressive peripheral sensory loss associated with progressive hearing impairment and early-onset dementia.

Specificity

This rabbit polyclonal antibody specifically detects DNA (cytosine-5)-methyltransferase 1 acetylated on four lysine residues from the C-terminal half containing six tandem repeats of K-G.

Immunogen

KLH-conjugated linear peptide corresponding to 11 amino acids from the C-terminal half with acetylated K1111, 1113, 1115, and 1117.

Application

Anti-DNMT1, Ac-4K, Cat. No. ABE1956, is a highly specific rabbit polyclonal antibody that targets DNA (cytosine-5)-methyltransferase 1 (DNMT1) and has been tested for use in Dot Blot, Immunocytochemistry, and Western Blotting.
Research Category
Epigenetics & Nuclear Function
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected DNA (cytosine-5)-methyltransferase 1 (DNMT1) in 293T cells transfected with pcmv-Tag2B-DNMT1 (Courtesy of an Independent Researcher).

Immunocytochemistry Analysis: A representative lot detected DNA (cytosine-5)-methyltransferase 1 (DNMT1) in Immunocytochemistry applications (Cheng, J., et. al. (2015). Nat Commun. 6:7023).

Dot Blot Analsysi: A representative lot detected DNA (cytosine-5)-methyltransferase 1 (DNMT1) in Dot Blot applications (Cheng, J., et. al. (2015). Nat Commun. 6:7023).

Western Blotting Analysis: A representative lot detected DNA (cytosine-5)-methyltransferase 1 (DNMT1) in Western Blotting applications (Cheng, J., et. al. (2015). Nat Commun. 6:7023).

Quality

Evaluated by Dot Blot in DNMT1-Control peptide & DNMT1-4Ac peptide.

Dot Blot Analysis: A 1:500 dilution of this antibody detected DNA (cytosine-5)-methyltransferase 1 with acetylated Lysine 1111, 1113, 1115, and 1117.

Target description

~200 kDa observed; 183.16 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Format: Purified
Purified rabbit polyclonal antibody in PBS with 0.02% sodium azide and 50% glycerol.

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service