Skip to Content
Merck
All Photos(1)

Documents

71059-M

Sigma-Aldrich

RosettaBlue(DE3) Competent Cells - Novagen

Escherichia coli, rod shaped

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106202
NACRES:
NA.81

product name

RosettaBlue(DE3) Competent Cells - Novagen, RosettaBlue host strains are NovaBlue derivatives that combine high transformation efficiency and with enhanced expression of eukaryotic proteins that contain codons rarely used in E. coli.

biological source

Escherichia coli

Quality Level

manufacturer/tradename

Novagen®

storage condition

OK to freeze

growth mode

adherent or suspension

morphology

rod shaped

technique(s)

microbiological culture: suitable

cell transformation

transformation efficiency: >1×108 cfu/μg

shipped in

dry ice

storage temp.

−70°C

General description

Genotype: endA1 hsdR17 (rK12 mK12+) supE44 thi-1 recA1 gyrA96 relA1 lac (DE3) F′[proA+B+ lacIqZΔM15::Tn10] pRARE (CamR, TetR)





This product contains genetically modified organisms (GMO). Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively. This legislation obliges us to request certain information about you and the establishment where the GMOs are being handled. Click here for Enduser Declaration (EUD) Form.
RosettaBlue host strains are NovaBlue derivatives that combine high transformation efficiency and recA endAlacIq mutations with enhanced expression of eukaryotic proteins that contain codons rarely used in E. coli. These strains supply tRNAs for AGG, AGA, AUA, CUA, CCC, and GGA on a chloramphenicol-resistant plasmid. The tRNA genes are driven by their native promoters. Blue/white screening is not possible with RosettaBlue(DE3) strains due to the presence of the lacZ α-peptide coding sequence in the λDE3 prophage.

DE3 indicates that the host is a lysogen of λDE3, and therefore carries a chromosomal copy of the T7 RNA polymerase gene under control of the lacUV5 promoter. Such strains are suitable for production of protein from target genes cloned in pET vectors by induction with IPTG.
RosettaBlue host strains are NovaBlue derivatives that combine high transformation efficiency and with enhanced expression of eukaryotic proteins that contain codons rarely used in E. coli.

Components

0.4 ml1 mlComponent

•2 × 0.2 ml5 × 0.2 mlRosettaBlue(DE3) Competent Cells

•2 × 2 ml4 × 2 mlSOC Medium

•2 ng2 ngTest Plasmid

Warning

Toxicity: Multiple Toxicity Values, refer to MSDS (O)

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service