Skip to Content
Merck
All Photos(1)

Key Documents

5.04815

Sigma-Aldrich

Salmeterol Xinafoate

Synonym(s):

Salmeterol Xinafoate, (±) 4-Hydroxy-a1-[[[6-(4-phenylbutoxy)hexyl]amino]m-ethyl]-1,3-benzenedimethanol xinafoate, GR 33343X xinafoate, β₂-Adrenoceptor Agonist, Salmeterol Xinafoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H37NO4 · C11H8O3
CAS Number:
Molecular Weight:
603.75
MDL number:
UNSPSC Code:
12352200

Assay

≥98% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

storage temp.

10-30°C

InChI

1S/C25H37NO4.C11H8O3/c27-20-23-18-22(13-14-24(23)28)25(29)19-26-15-7-1-2-8-16-30-17-9-6-12-21-10-4-3-5-11-21;12-10-8-4-2-1-3-7(8)5-6-9(10)11(13)14/h3-5,10-11,13-14,18,25-29H,1-2,6-9,12,15-17,19-20H2;1-6,12H,(H,13,14)

InChI key

XTZNCVSCVHTPAI-UHFFFAOYSA-N

General description

A potent and selective β2-adrenoceptor agonist (EC50 = 5.3 nM) that displays bronchodilator actions. Unlike other β2- adrenoceptor agonists, Salmeterol binds to exo-site domain of β2-AR receptors, producing a slow onset of action and prolonged activation. Clinically used for the treatment of asthma and chronic obstructive pulmonary disease(COPD).
A potent and selective ?2-adrenoceptor agonist (EC50 = 5.3 nM) that displays bronchodilator actions. Unlike other ?2- adrenoceptor agonists, Salmeterol binds to exo-site domain of ?2-AR receptors, producing a slow onset of action and prolonged activation. Clinically used for the treatment of asthma and chronic obstructive pulmonary disease(COPD).

Biochem/physiol Actions

Primary Target
→₂-adrenoceptor

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Roberts, J., et al. 1999. Eur. Respir. J.14, 275.
Rennard, S., et al. 2001. Am. J. Respir. Crit. Care Med.163, 1087.
Akabane, H., et al. 2006. Allergol. Int.55, 387.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service