Skip to Content
Merck
All Photos(1)

Documents

219011

Sigma-Aldrich

Caspase Inhibitor VI

InSolution, ≥95%

Synonym(s):

InSolution Caspase Inhibitor VI

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H28FN3O7
Molecular Weight:
453.46
UNSPSC Code:
12352200
NACRES:
NA.54

Quality Level

Assay

≥95% (TLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated
protect from light

shipped in

wet ice

storage temp.

−20°C

General description

A cell-permeable, irreversible, and general caspase inhibitor. Also reported to inhibit Peptide: N-glycanase (PNGase) in vitro and in vivo. Useful for studies involving recombinant, isolated, and purified caspase enzymes. Unlike Caspase Inhibitor I (Cat. No. 627610), this inhibitor does not require pretreatment with esterase for in vitro studies.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
general caspase inhibitor
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Sequence

Z-Val-Ala-Asp-CH₂F*

Physical form

A 10 mM (1 mg/221 µl) solution of Caspase Inhibitor VI (Cat. No. 219007) in DMSO.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C). Aliquots are stable for up to 3 months at -20°C.

Other Notes

Misaghi, S., et al. 2004. Chem. Biol.11, 1677.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F

Flash Point(C)

87 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service