Skip to Content
Merck
All Photos(1)

Key Documents

1951-M

Millipore

Bacitracin

Synonym(s):

Bacitracin, Altracin, PDI Inhibitor I, Protein Disulfide Isomerase Inhibitor I

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200

form

solid

Quality Level

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

gray-white

solubility

ethanol: soluble
water: soluble

storage temp.

2-8°C

InChI

1S/C66H103N17O16S.Zn/c1-9-35(6)52(69)66-81-48(32-100-66)63(97)76-43(26-34(4)5)59(93)74-42(22-23-50(85)86)58(92)83-53(36(7)10-2)64(98)75-40-20-15-16-25-71-55(89)46(29-49(68)84)78-62(96)47(30-51(87)88)79-61(95)45(28-39-31-70-33-72-39)77-60(94)44(27-38-18-13-12-14-19-38)80-65(99)54(37(8)11-3)82-57(91)41(21-17-24-67)73-56(40)90;/h12-14,18-19,31,33-37,39-48,52-54H,9-11,15-17,20-30,32,67,69H2,1-8H3,(H2,68,84)(H,71,89)(H,73,90)(H,74,93)(H,75,98)(H,76,97)(H,77,94)(H,78,96)(H,79,95)(H,80,99)(H,82,91)(H,83,92)(H,85,86)(H,87,88);

InChI key

QSNOBVJFKSQBBD-UHFFFAOYSA-N

General description

A polypeptide antibiotic and a peptidase inhibitor. Active against gram-positive microorganisms. An inhibitor of protein disulfide isomerase. Resembles penicillin in causing protoplast formation and inhibition of cell wall synthesis. Blocks the regeneration of the carrier phospholipid. Stimulates 5-hydroxytryptamine receptors and increases vascular resistance. Bacitracin is unstable in alkaline solutions. Note: Do not use a glass rod for stirring the solution, it may cause some precipitation.
A polypeptide antibiotic and peptidase inhibitor. An inhibitor of protein disulfide isomerase. Resembles penicillin in causing protoplast formation and inhibition of cell wall synthesis. Blocks the regeneration of carrier phospholipid. Stimulates 5-hydroxytryptamine receptors and increases vascular resistance.
Note: 1 KU = 1000 units.

Biochem/physiol Actions

Potency ≥40 Bacitracin units/mg solid, as is

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of antibiotic equal to 26 µg of the USP standard. Note: 1 ku = 1000 units.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 2 months at -20°C.

Other Notes

Ikai, Y., et al. 1995. J. Antibiot. (Tokyo) 48, 233.
Clive, D.R., et al. 1994. Exp. Cell Res. 214, 139.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service