Skip to Content
Merck
All Photos(1)

Documents

1.18465

Supelco

Phosphate Test Refill pack

for use with MCOLORTEST®

Synonym(s):

Phosphate Visual Test Kit Refills

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

feature

refill

Quality Level

specific analyte(s)

phosphate

compatibility

for use with MCOLORTEST®

detection method

colorimetric

storage temp.

15-25°C

General description

This refill pack contains all the reagents for the MQuant® test kits 1.14846 and 1.18394 for the colorimetric evaluation of phosphate. When the reagents of the MQuant® test are used up, the kit can be replenished by ordering the appropriate refill pack. Refill packs are available for a range of MQuant® test kits, making it a particularly economical and ecological system.

Method
In sulfuric solution orthophosphate ions react with molybdate ions to form molybdophosphoric acid. Ascorbic acid reduces this to phosphomolybdenum blue (PMB). The phosphate concentration is measured semiquantitatively by visual colorimetric evaluation.

Legal Information

MCOLORTEST is a registered trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service