MAT239353084
Matrix Scientific
2-Amino-1-benzyl-4,5-diphenyl-1H-pyrrole-3-carbonitrile
About This Item
mp
175-179
Iupac Name
2-amino-1-benzyl-4,5-diphenyl-1H-pyrrole-3-carbonitrile
Inchi Code
1S/C24H19N3/c25-16-21-22(19-12-6-2-7-13-19)23(20-14-8-3-9-15-20)27(24(21)26)17-18-10-4-1-5-11-18/h1-15H,17,26H2
InChI key
GXIMOGHICAZEKA-UHFFFAOYSA-N
Purity
Country of Origin
Product Link
Melting Point
MSDS
https://www.matrixscientific.com/045859.html
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.