Skip to Content
Merck
All Photos(1)

Key Documents

KEY371462929

Key Organics / BIONET

2-amino-N-methyl-2-phenylacetamide

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
164.21
MDL number:

Iupac Name

2-amino-N-methyl-2-phenylacetamide

Inchi Code

1S/C9H12N2O/c1-11-9(12)8(10)7-5-3-2-4-6-7/h2-6,8H,10H2,1H3,(H,11,12)

InChI key

YEKJABDQUZLZOA-UHFFFAOYSA-N

Purity

90%

Country of Origin

GB

Physical Form

Liquid

Product Link

https://www.keyorganics.net/sku/SS-3472

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

MSDS

https://www.keyorganics.net/sds/SS-3472


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library