Skip to Content
Merck
All Photos(1)

Key Documents

KEY298198565

Key Organics / BIONET

2-chloro-6-(furan-2-yl)pyridine-3-carbonitrile

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
204.62
MDL number:

mp

130 - 132

Iupac Name

2-chloro-6-(2-furyl)nicotinonitrile

Inchi Code

1S/C10H5ClN2O/c11-10-7(6-12)3-4-8(13-10)9-2-1-5-14-9/h1-5H

InChI key

HMPRWDLTTCZMAY-UHFFFAOYSA-N

Purity

95%

Country of Origin

GB

Physical Form

Solid

Product Link

https://www.keyorganics.net/sku/FE-0767

Melting Point

130 - 132

Pictograms

Exclamation mark

Signal Word

Warning

MSDS

https://www.keyorganics.net/sds/FE-0767


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library