Skip to Content
Merck
All Photos(1)

Key Documents

ENA447956939

Enamine

5-phenyl-1,2,3,6-tetrahydropyridine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
195.69
MDL number:

mp

210-214

Iupac Name

5-phenyl-1,2,3,6-tetrahydropyridine hydrochloride

Inchi Code

1S/C11H13N.ClH/c1-2-5-10(6-3-1)11-7-4-8-12-9-11;/h1-3,5-7,12H,4,8-9H2;1H

InChI key

AERNNJNTNZDJDC-UHFFFAOYSA-N

storage temp.

RT

Salt Data

Cl

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-206389

Melting Point

210-214

Pictograms

Exclamation mark

Signal Word

Warning

MSDS

https://www.enaminestore.com/catalog/EN300-206389


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library