COM448633873
Combi-Blocks, Inc.
3-Chloro-2-(N,N-diethylaminomethyl)phenylboronic acid, pinacol ester
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
N-[2-chloro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]-N-ethylethanamine
Inchi Code
1S/C17H27BClNO2/c1-7-20(8-2)12-13-14(10-9-11-15(13)19)18-21-16(3,4)17(5,6)22-18/h9-11H,7-8,12H2,1-6H3
InChI key
IMTUNIYOSPVFSO-UHFFFAOYSA-N
storage temp.
Refrigerated
Purity
98%
Country of Origin
CN
Product Link
https://www.combi-blocks.com/cgi-bin/find.cgi?PN-4246
MSDS
https://www.combi-blocks.com/cgi-bin/find.cgi?PN-4246
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.