COM448619554
Combi-Blocks, Inc.
4-Bromo-N-methylaniline hydrochloride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
N-(4-bromophenyl)-N-methylamine hydrochloride
Inchi Code
1S/C7H8BrN.ClH/c1-9-7-4-2-6(8)3-5-7;/h2-5,9H,1H3;1H
InChI key
LFVNPPHDWREZHC-UHFFFAOYSA-N
Purity
98%
Country of Origin
CN
Product Link
https://www.combi-blocks.com/cgi-bin/find.cgi?HC-2800
MSDS
https://www.combi-blocks.com/cgi-bin/find.cgi?HC-2800
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.