Skip to Content
Merck
All Photos(1)

Key Documents

C12H316CE9B6

Chemcia Scientific, LLC

3-Cyclopropoxy-benzonitrile

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
159.19
MDL number:
Pricing and availability is not currently available.

Iupac Name

3-(cyclopropyloxy)benzonitrile

Inchi Code

1S/C10H9NO/c11-7-8-2-1-3-10(6-8)12-9-4-5-9/h1-3,6,9H,4-5H2

InChI key

NVBGCWUMVIHESK-UHFFFAOYSA-N

storage temp.

0-5C

Purity

90%

Country of Origin

US

Shipping Temperature

Ambient

Physical Form

Yellow Liquid

Product Link

http://www.chemcia.com/product_fl02.asp?id=2628

MSDS

http://www.chemcia.com/product_fl02.asp?id=2628


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library