C12H316CE30A
Chemcia Scientific, LLC
4-Iodo-3-methoxy-5-nitrobenzaldehyde
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
4-iodo-3-methoxy-5-nitrobenzaldehyde
Inchi Code
1S/C8H6INO4/c1-14-7-3-5(4-11)2-6(8(7)9)10(12)13/h2-4H,1H3
InChI key
WVDDJASTMZNUNH-UHFFFAOYSA-N
storage temp.
0-5C
Purity
97%
Country of Origin
US
Shipping Temperature
Ambient
Physical Form
Light Yellow Solid
Product Link
http://www.chemcia.com/product_fl02.asp?id=251
MSDS
http://www.chemcia.com/product_fl02.asp?id=251
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.