Skip to Content
Merck
All Photos(1)

Key Documents

CH4424847616

ChemBridge Corporation

4-(4-piperidinyloxy)benzoic acid hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
257.72
MDL number:
Pricing and availability is not currently available.

Iupac Name

4-(piperidin-4-yloxy)benzoic acid hydrochloride

Inchi Code

1S/C12H15NO3.ClH/c14-12(15)9-1-3-10(4-2-9)16-11-5-7-13-8-6-11;/h1-4,11,13H,5-8H2,(H,14,15);1H

InChI key

NHTKIPKHDVKBJY-UHFFFAOYSA-N

storage temp.

RT

Salt Data

HCl

Purity

95%

Country of Origin

KZ

Shipping Temperature

RT

Physical Form

solid

Product Link

https://www.hit2lead.com/building-blocks/4039677

MSDS

https://www.hit2lead.com/sds/4039677_sds.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library