Skip to Content
Merck
All Photos(1)

Key Documents

ATEH984B2ED9

AstaTech, Inc.

{3-PHENYLBICYCLO[1.1.1]PENTAN-1-YLMETHANOL

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
174.24
MDL number:

Iupac Name

(3-phenylbicyclo[1.1.1]pentan-1-yl)methanol

Inchi Code

1S/C12H14O/c13-9-11-6-12(7-11,8-11)10-4-2-1-3-5-10/h1-5,13H,6-9H2

InChI key

FPXQMBITZDVAOP-UHFFFAOYSA-N

Purity

95%

Country of Origin

CN

Physical Form

Pale-yellow to Yellow-brown Liquid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=177108

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=P20050


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library