Skip to Content
Merck
All Photos(1)

Key Documents

ATEH57C9A1E8

AstaTech, Inc.

5-BROMO-1,2,3,4-TETRAHYDRO-1,8-NAPHTHYRIDINE

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
213.08
MDL number:

Iupac Name

5-bromo-1,2,3,4-tetrahydro[1,8]naphthyridine

Inchi Code

1S/C8H9BrN2/c9-7-3-5-11-8-6(7)2-1-4-10-8/h3,5H,1-2,4H2,(H,10,11)

InChI key

CYTUKWQXFDIKLS-UHFFFAOYSA-N

Purity

95%

Country of Origin

CN

Physical Form

Pale-yellow to Yellow-brown Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=123882

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=W14857


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library