ATE959439867
AstaTech, Inc.
5-METHYL-5,6-DIHYDRO-4H-PYRROLO[1,2-B]PYRAZOLE
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
5-methyl-5,6-dihydro-4H-pyrrolo[1,2-b]pyrazole
Inchi Code
1S/C7H10N2/c1-6-4-7-2-3-8-9(7)5-6/h2-3,6H,4-5H2,1H3
InChI key
LRDQMNHYYHKGTO-UHFFFAOYSA-N
Purity
95%
Country of Origin
CN
Physical Form
Yellow to Brown Sticky Oil to Semi-Solid
Product Link
http://astatechinc.com/CPSResult.php?CRNO=110526
MSDS
http://astatechinc.com/Asta_MSDS.php?Catalog=56875
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.