Documents
Z648450
Quickfit® bends with vacuum connection
socket joint: ST/NS 24/29, cone joint: ST/NS 24/29
Synonym(s):
RA3/33/SC
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
Joint
cone joint: ST/NS24/29
socket joint: ST/NS24/29
manufacturer/tradename
Quickfit RA 3/33/SC
availability
not available in USA and Canada
not available in USA and Canada
General description
Socket to cone. With screw-thread adapter for flexible tubing.
Legal Information
Quickfit is a registered trademark of SciLabware Limited