Z544477
Molymod® molecular model set
teacher/researcher set, complex ions
About This Item
Recommended Products
feature
no. of atoms 156
no. of bonds 160
manufacturer/tradename
Molymod MMS-006
General description
(6) Red 2-hole angular oxygen atom parts
(20) White single-hole hydrogen atom parts
(4) Blue nitrogen atom parts
(2) Yellow sulfur atom parts: (1) tetrahedral, (1) octahedral
(1) Purple tetrahedral phosphorus atom part
(4) Green single-hole halogen atom parts
(1) Grey single-hole metal atom parts
(26) Medium-length 19mm grey links
(12) Flexible long 30mm grey links
(26) Short 2mm white links
Link remover tool
Storage box
Instructions
Scale is 2.8 cm =100 pm (open style) and 1.45 cm = 100 pm (compact, semi-space-filling). Sets include standard color-coded plastic atom centers (with bond holes set at the appropriate angles), bonding links, and a plastic storage box.
Set pictured is representative of the Student set.
See our Selection Guide for detailed descriptions of set contents.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)