Skip to Content
Merck
All Photos(1)

Documents

THS04147

ThalesNano MicroCatCart catalyst cartridge system

L × diam. 30 mm × 4 mm, 1% Pt/Al2O3

Synonym(s):

Heterogeneous catalysis system, safe Heterogeneous catalysis

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41103817
NACRES:
NB.51

packaging

pkg of 6 ea

manufacturer/tradename

ThalesNano THS 04147

L × diam.

30 mm × 4 mm

Looking for similar products? Visit Product Comparison Guide

Related Categories

Legal Information

MicroCatCart is a trademark of ThalesNano Nanotechnology Inc.

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service