MT51107690
Mettler Toledo DX series ISE half-cell membrane kit
For Fluoroborate ISE
All Photos(1)
About This Item
manufacturer/tradename
Mettler-Toledo 51107690
General description
DX series ion specific membrane kit. Mount the new membrane onto the shaft of your ISE half-cell, and you have a new ISE!
Kit contains one membrane plus electrolyte
Kit contains one membrane plus electrolyte