Skip to Content
Merck
All Photos(2)

Documents

L4206

Sigma-Aldrich

3,5-Lutidine

≥98%

Synonym(s):

3,5-Dimethylpyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H9N
CAS Number:
Molecular Weight:
107.15
Beilstein:
105682
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

1.5 mmHg ( 20 °C)

Assay

≥98%

form

liquid

refractive index

n20/D 1.504 (lit.)

bp

169-170 °C (lit.)

mp

−9 °C (lit.)

density

0.939 g/mL at 25 °C (lit.)

SMILES string

Cc1cncc(C)c1

InChI

1S/C7H9N/c1-6-3-7(2)5-8-4-6/h3-5H,1-2H3

InChI key

HWWYDZCSSYKIAD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

116.6 °F - closed cup

Flash Point(C)

47 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael Wleklinski et al.
Physical chemistry chemical physics : PCCP, 17(28), 18364-18373 (2015-06-25)
Electrospray ionization of metal salt solutions followed by ambient heating transforms the resulting salt clusters into new species, primarily naked ionic metal clusters. The experiment is done by passing the clusters through a heated coiled loop outside the mass spectrometer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service