Skip to Content
Merck
All Photos(1)

Key Documents

C19201

Sigma-Aldrich

Chloroacetaldehyde diethyl acetal

99%

Synonym(s):

2-Chloro-1,1-diethoxyethane, Chloroacetal

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CH(OC2H5)2
CAS Number:
Molecular Weight:
152.62
Beilstein:
969254
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

<0.02 atm ( 20 °C)

Quality Level

Assay

99%

form

liquid

autoignition temp.

401 °F

refractive index

n20/D 1.416 (lit.)

bp

157 °C (lit.)

density

1.018 g/mL at 25 °C (lit.)

SMILES string

CCOC(CCl)OCC

InChI

1S/C6H13ClO2/c1-3-8-6(5-7)9-4-2/h6H,3-5H2,1-2H3

InChI key

OVXJWSYBABKZMD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

84.2 °F

Flash Point(C)

29 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J C Kandala et al.
Biochemical and biophysical research communications, 167(2), 457-463 (1990-03-16)
The effect of 2-chloroacetaldehyde, CAA, a metabolite of vinyl chloride and 2-chloroacetal, CAC, an ethyl diester of chloroacetaldehyde, on DNA synthesis in animal cells has been investigated. Both compounds drastically inhibited DNA synthesis at 10 to 20 microM. The inhibitory

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service