Skip to Content
Merck
All Photos(1)

Documents

BR781641

BRAND® microplate BRANDplates®, pureGrade

size 1536 wells, size 10 μL, flat bottom, white

Synonym(s):

BRANDplates®, Microplates, medium binding, pureGrade

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.22

material

flat bottom
polystyrene
white wells

sterility

non-sterile

feature

lid: no
skirt (F-bottom)

packaging

pack of 50 ea

manufacturer/tradename

BRAND 781641

size

10 μL
1536 wells

working volume

10 μL

color

white

binding type

medium binding surface

Looking for similar products? Visit Product Comparison Guide

General description

The BRAND® Microplate BRANDplates® PureGrade features a medium-binding, untreated surface, making it an ideal choice for a variety of applications. This non-sterile microplate serves as the standard for use in homogeneous assays, screenings, and sample storage. It is non-cytotoxic, making it safe for use in cell-based assays. With BIO-CERT® CERTIFIED QUALITY, this microplate is guaranteed to be free from human DNA, RNase, DNase, and pyrogens. This versatile microplate is a dependable option for researchers looking for consistent performance across numerous laboratory applications.

Features and Benefits

• Optimal surfaces for optimal results• Reliable quality from the cleanroom• Tested by independent laboratories

Legal Information

BIO-CERT is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
BRANDplates is a registered trademark of BRAND GMBH + CO KG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service