Key Documents
BAF420741226
SP Bel-Art® Secador® 4.0 Vertical Auto-Desiccator Cabinet
internal volume 1.9 ft3, AC/DC input 220 V, universal plug set, blue cap (with blue end caps)
Synonym(s):
230 V, 4.0 VERTICAL AUTO-DESICCATOR CABINETS WITH BLUE END-CAPS, SECADOR 4.0 VERTICAL AUTO-DESICCATOR CABINETS, SP BEL-ART SECADOR VERTICAL PROFILE CLEAR 4.0 AUTO-DESICCATOR CABINET WITH BLUE END CAPS, SP BEL-ART SECADOR VERTICAL PROFILE CLEAR 4.0 AUTO-DESICCATOR CABINET WITH BLUE END CAPS; 230V, 1.9 CU. FT.
About This Item
UNSPSC Code:
41104510
material
blue cap (with blue end caps)
clear polyester (Co-polyester)
manufacturer/tradename
Bel-Art F42074-1226
internal volume
1.9 ft3
General description
Protecting valuable humidity-sensitive items, such as reagents or electronics, is easy and fully automatic with Secador® Auto-Desiccator Cabinets. The patented, electronic desiccant regeneration module recharges the desiccant every 20 minutes. Compared to other auto-desiccators that regenerate once every 4 to 6 hours, the Secador® Auto-Desiccator quickly attains a low %rH level on start-up and consistently maintains low %rH, especially with frequent entry to the desiccator interior.
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Secador is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)