Skip to Content
Merck
All Photos(1)

Key Documents

742813

Sigma-Aldrich

(S)-2,2-Dimethyl-1-(1-naphthyl)propylamine

≥98% (HPLC)

Synonym(s):

S)-α-(1,1-Dimethylethyl)-1-naphthalenemethanamine, (S)-α-tert-Butyl-1-naphthalenemethanamine, (S)-α-tert-Butyl-1-naphthylmethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H19N
CAS Number:
Molecular Weight:
213.32
Beilstein:
8684336
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -59.0±3°, c = 0.1 in dichloromethane

storage temp.

2-8°C

SMILES string

N[C@@H](C(C)(C)C)C1=CC=CC2=C1C=CC=C2

InChI

1S/C15H19N/c1-15(2,3)14(16)13-10-6-8-11-7-4-5-9-12(11)13/h4-10,14H,16H2,1-3H3/t14-/m1/s1

InChI key

FQTPRCXAYUOOGA-CQSZACIVSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service